To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Dopamine
- Antigen Symbol:Dopamine
- Clonality:Monoclonal
- Clone:2B11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:0.2 mg
- Environmentally Preferable:
- Antigen Synonyms:DA|3 hydroxytyramine|Oxytyramine|Dopamine|3 4 dihydroxyphenethylamine
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Chemical/ Small Molecule conjugated to BSA.
- Purification:Protein A or G purified
- Cat. No.:102121-648
- Supplier no.:NB110-2538
The Dopamine Antibody (2B11) from Novus Biologicals is a mouse monoclonal antibody to Dopamine. This antibody reacts with human. The Dopamine Antibody (2B11) has been validated for the following applications: Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Dopamine
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human