To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome P450 1A1/1A2
- Antigen Symbol:CYP1A1/1A2
- Clonality:Monoclonal
- Clone:MC1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Rat,Mouse
- Western Blot:Yes
- Size:100μl
- Environmentally Preferable:
- Gene ID:1543
- Antigen Synonyms:AHRR|subfamily A|Cytochrome P450-P1|CYP1|cytochrome P450|CYPIA1|EC 1.14.14.1|Cytochrome P450-C|subfamily I (aromatic compound-inducible)|cytochrome P450 1A1|Cytochrome P450 form 6|AHH|polypeptide 1|family 1|P450DX|cytochrome P1-450|xenobiotic monooxygenase|aryl hydrocarbon hydroxylase|dioxin-inducible|flavoprotein-linked monooxygenase|P450-C|CP11|P1-450
- Storage Buffer:Ascites
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:3-methylcholanthrene induced rat cytochrome P450 protein.
- Purification:Unpurified
- Cat. No.:102116-318
- Supplier no.:NB100-74398
Specifications
About this item
The Cytochrome P450 1A1 / 1A2 Antibody (MC1) from Novus Biologicals is a mouse monoclonal antibody to Cytochrome P450 1A1 / 1A2. This antibody reacts with human, mouse, rat, primate. The Cytochrome P450 1A1 / 1A2 Antibody (MC1) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Cytochrome P450 1A1/1A2
Clonality: Monoclonal
Clone: MC1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate