To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Mindin/Spondin-2
- Antigen Symbol:SPON2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Epitope:C-RKNQYVSNGLRDFA
- Gene ID:10417
- Antigen Synonyms:MINDIN|DIL1FLJ34460|spondin-2|FLJ16313|DIL-1|DKFZp686G21139|M-SPONDIN|Differentially expressed in cancerous and non-cancerous lung cells 1|extracellular matrix protein|spondin 2|FLJ22401
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-RKNQYVSNGLRDFA corresponding to internal region according to NP_036577.1.
- Purification:Immunogen affinity purified
- Cat. No.:102182-550
- Supplier no.:NBP1-52031
Specifications
About this item
Goat Polyclonal Spondin-2 Antibody. Tested Applications: Western Blot, Peptide ELISA. Tested Reactivity: Human, Mouse, Rat, Bovine.
Type: Primary
Antigen: SPON2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human