To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Protein Kinase C Delta Binding Protein
- Antigen Symbol:PRKCDBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:112464
- Antigen Synonyms:hSRBC|protein kinase C|Cavin-3|MGC20400|delta binding protein|SRBCprotein kinase C delta-binding protein|sdr-related gene product that binds to c-kinase|Serum deprivation response factor-related gene product that binds to C-kinase
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-RRTGLQKVQSLR corresponding to internal region according to NP_659477.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102174-650
- Supplier no.:NBP1-45210
The PRKCDBP Antibody from Novus Biologicals is a goat polyclonal antibody to PRKCDBP. This antibody reacts with human, mouse, rat. The PRKCDBP Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: PRKCDBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat