To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Aldehyde Dehydrogenase 5 Family, member A1
- Antigen Symbol:ALDH5A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Epitope:C-TGKILLHHAANS
- Gene ID:7915
- Antigen Synonyms:EC 1.2.1.24|member A1|EC 1.2.1|aldehyde dehydrogenase 5 family|SSADHmitochondrial|NAD(+)-dependent succinic semialdehyde dehydrogenase
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Peptide with sequence C-TGKILLHHAANS corresponding to internal region according to NP_001071.1.
- Purification:Immunogen affinity purified
- Cat. No.:102182-670
- Supplier no.:NBP1-52100
The ALDH5A1 Antibody from Novus Biologicals is a goat polyclonal antibody to ALDH5A1. This antibody reacts with human, mouse, rat. The ALDH5A1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Peptide
Type: Primary
Antigen: ALDH5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-TGKILLHHAANS
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat