To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Tyrosine Phosphatase Receptor Type D2
- Antigen Symbol:PTPN14/PTPD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:5784
- Antigen Synonyms:tyrosine-protein phosphatase non-receptor type 14|Protein-tyrosine phosphatase pez|non-receptor type 14|PEZcytoskeletal-associated protein tyrosine phosphatase|protein tyrosine phosphatase|EC 3.1.3|MGC126803|PTP36|EC 3.1.3.48
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:The immunogen for this antibody was a sequence from 580a.a. to 848a.a. from PTPD2(a 1187a.a. long protein)
- Purification:Immunogen affinity purified
- Cat. No.:102116-694
- Supplier no.:NB100-75310
Specifications
About this item
The PTPN14 / PTPD2 Antibody from Novus Biologicals is a chicken polyclonal antibody to PTPN14 / PTPD2. This antibody reacts with human, mouse. The PTPN14 / PTPD2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PTPN14/PTPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse