To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin-conjugating Enzyme E2 Variant 2
- Antigen Symbol:UEV2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:7336
- Antigen Synonyms:S. cerevisiae|UEV2|DDVit-1|DDVIT1|1 alpha|Vitamin D3-inducible protein|MMS2MMS2 homolog|EDPF1|ubiquitin-conjugating enzyme E2 variant 2|25-dihydroxyvitamin D3-inducible|EDAF-1|UEV-2|methyl methanesulfonate sensitive 2|EDPF-1DDVit 1|enterocyte differentiation-associated factor EDAF-1|Enterocyte differentiation-promoting factor 1|enterocyte differentiation promoting factor|Enterocyte differentiation-associated factor 1|homolog of
- Storage Buffer:PBS containing 0.2% gelatin
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:This antibody was generated by immunizing rabbit with a mixture of synthetic peptides containing amino acids 12-28 of human UBE2V2.
- Purification:Protein G purified
- Cat. No.:102109-932
- Supplier no.:NB100-56360
Specifications
About this item
The UEV2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UEV2. This antibody reacts with human. The UEV2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: UEV2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human