About this item
The NSP 5 alpha 3 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to NSP 5 alpha 3 alpha. This antibody reacts with human. The NSP 5 alpha 3 alpha Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: NSP 5 alpha 3 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102109-430
- 102109-444
- Antigen Symbol:
- NSP 5 alpha 3 alpha
- NSP 5 alpha 3 alpha
- Antigen Name:
- NSP 5 alpha 3 alpha
- NSP 5 alpha 3 alpha
- Antigen Synonyms:
- FLJ36955|cytospin-B|sperm antigen with calponin-like and coiled coil domains 1|structure protein NSP5a3a|structure protein NSP5b3a|CYTSBcytospin B|Nuclear structure protein 5|sperm antigen with calponin homology and coiled coil domains 1|structure protein NSP5b3b|NSP|NSP5|sperm antigen HCMOGT 1|structure protein NSP5a3b|sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B|Sperm antigen HCMOGT-1|HCMOGT-1
- FLJ36955|cytospin-B|sperm antigen with calponin-like and coiled coil domains 1|structure protein NSP5a3a|structure protein NSP5b3a|CYTSBcytospin B|Nuclear structure protein 5|sperm antigen with calponin homology and coiled coil domains 1|structure protein NSP5b3b|NSP|NSP5|sperm antigen HCMOGT 1|structure protein NSP5a3b|sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B|Sperm antigen HCMOGT-1|HCMOGT-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 92521
- 92521
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 1-100). [UniProt# Q5M775]
- An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 1-100). [UniProt# Q5M775]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:
- Store at 4C. Do not freeze.
- Store at 4C. Do not freeze.