About this item
Rabbit Polyclonal IRE1 alpha Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Primate (-).
Type: Primary
Antigen: IRE1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102107-140
- 102107-126
- Antigen Symbol:
- IRE1A
- IRE1A
- Antigen Name:
- IRE1 alpha
- IRE1 alpha
- Antigen Synonyms:
- IRE 1a|Endoplasmic reticulum-to-nucleus signaling 1|ER to nucleus signalling 1|Inositol-requiring protein 1|inositol-requiring 1|hIRE1p|protein kinase/endoribonuclease|endoplasmic reticulum to nucleus signaling 1|Ire1-alpha|IRE1P|IRE1MGC163279|EC 2.7.11|serine/threonine-protein kinase/endoribonuclease IRE1|IRE1a|MGC163277|FLJ30999
- IRE 1a|Endoplasmic reticulum-to-nucleus signaling 1|ER to nucleus signalling 1|Inositol-requiring protein 1|inositol-requiring 1|hIRE1p|protein kinase/endoribonuclease|endoplasmic reticulum to nucleus signaling 1|Ire1-alpha|IRE1P|IRE1MGC163279|EC 2.7.11|serine/threonine-protein kinase/endoribonuclease IRE1|IRE1a|MGC163277|FLJ30999
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2081
- 2081
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460]
- A synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Tris-Glycine and 0.15M NaCl
- Tris-Glycine and 0.15M NaCl
- Storage Temperature:
- Store at -20C. Avoid freeze-thaw cycles.
- Store at -20C. Avoid freeze-thaw cycles.