To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 4
- Antigen Symbol:GPR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Primates,Monkey,Human,Dog
- Size:0.05 ml
- Environmentally Preferable:
- Gene ID:2828
- Antigen Synonyms:G-protein coupled receptor 19|G-protein coupled receptor 4|G protein-coupled receptor 4|GPR19
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Synthetic 19 amino acid peptide from C-terminus of human GPR4.
- Purification:Immunogen affinity purified
- Cat. No.:102253-108
- Supplier no.:NLS145
Rabbit Polyclonal GPR4 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml to 24 ?g/ml
Type: Primary
Antigen: GPR4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human