To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Forkhead Box O3
- Antigen Symbol:FOXO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2309
- Antigen Synonyms:forkhead box protein O3|forkhead homolog (rhabdomyosarcoma) like 1|Drosophila|FKHRL1MGC31925|FKHRL1P2|AF6q21|MGC12739|DKFZp781A0677|forkhead box O3|forkhead|Forkhead in rhabdomyosarcoma-like 1|in rhabdomyosarcoma-like 1|FOXO3Aforkhead box O3A|FOXO2|homolog of|AF6q21 protein
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 625 and the C-terminus (residue 673) of human Forkhead Box O3a using the numbering given in Swiss-Prot entry O43524 (GeneID 2309).
- Purification:Immunogen affinity purified
- Cat. No.:102111-232
- Supplier no.:NB100-614
Specifications
About this item
The FOXO3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FOXO3. This antibody reacts with human, mouse. The FOXO3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FOXO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse