Order Entry
United States
ContactUsLinkComponent
Anti-pan Nuclear Pore Complex Marker Mouse Monoclonal Antibody [clone: 39C7]
Anti-pan Nuclear Pore Complex Marker Mouse Monoclonal Antibody [clone: 39C7]
Catalog # 102156-738
Supplier:  Novus Biologicals
Anti-pan Nuclear Pore Complex Marker Mouse Monoclonal Antibody [clone: 39C7]
Catalog # 102156-738
Supplier:  Novus Biologicals
Supplier Number:  NBP1-05390
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    pan Nuclear Pore Complex Marker
  • Antigen Symbol:
    pan Nuclear Pore Complex Marker
  • Clonality:
    Monoclonal
  • Clone:
    39C7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    0.5 ml
  • Environmentally Preferable:
  • Antigen Synonyms:
    Nuclear pore complex protein Nup107|Nucleoporin Nup107|NUP84|NPC proteins|NUP107|NUP 107|107 kDa nucleoporin|Nucleoporin 107kDa|NUP 84
  • Storage Buffer:
    No buffer
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Yeast nuclear preparations
  • Purification:
    Tissue culture supernatant
  • Cat. No.:
    102156-738
  • Supplier no.:
    NBP1-05390

Specifications

About this item

The pan Nuclear Pore Complex Marker Antibody (39C7) from Novus Biologicals is a mouse monoclonal antibody to pan Nuclear Pore Complex Marker. This antibody reacts with human, mouse, rat. The pan Nuclear Pore Complex Marker Antibody (39C7) has been validated for the following applications: Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: pan Nuclear Pore Complex Marker
Clonality: Monoclonal
Clone: 39C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat