To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD8 alpha
- Clonality:Monoclonal
- Clone:CT-8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Turkey,Birds,Chicken
- Size:0.5 mg
- Environmentally Preferable:
- Neutralisation:Yes
- Gene ID:925
- Antigen Synonyms:T8 T-cell antigen|p32|OKT8 T-cell antigen|T-cell antigen Leu2|RPA-T8|RPA-T8 Flow|alpha polypeptide (p32)|T-lymphocyte differentiation antigen T8/Leu-2|RPA-T8 antibody flow|Leu2 T-lymphocyte antigen|T-cell surface glycoprotein CD8 alpha chain|RPA-T8 Clone|T cell co-receptor|CD8 antigen|Leu2|MAL|CD8|RPA-T8 CD8|CD8a antigen|CD8a molecule
- Storage Buffer:0.1M BBS (pH 8.2)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.5 mg/ml
- Immunogen:Chicken thymocytes and Ig-negative blood leukocytes
- Purification:Protein A or G purified
- Cat. No.:102161-856
- Supplier no.:NBP1-28270
Specifications
About this item
The CD8 alpha Antibody (CT-8) from Novus Biologicals is a mouse monoclonal antibody to CD8 alpha. This antibody reacts with avian, chicken, turkey. The CD8 alpha Antibody (CT-8) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Blocking / Neutralizing, Immunocytochemistry.
Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: CT-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Avian, Chicken, Turkey