To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Mitotic checkpoint serine/threonine-protein kinase BUB1
- Antigen Symbol:BubR1
- Clonality:Monoclonal
- Clone:8G1
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:701
- Antigen Synonyms:Mitotic checkpoint kinase MAD3L|EC 2.7.11.1|MAD3/BUB1-related protein kinase|beta|hBUBR1|SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta|Protein SSK1|BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog)|budding uninhibited by benzimidazoles 1 homolog beta (yeast)|MAD3LBUB1beta|Bub1A
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A portion of human BubR1 tagged at the N-terminus with pMAL (residues 1-350). [UniProt# O60566]
- Purification:Protein G purified
- Cat. No.:102108-602
- Supplier no.:NB100-353B
The BubR1 Antibody (8G1) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to BubR1. This antibody reacts with human, mouse. The BubR1 Antibody (8G1) [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: BubR1
Clonality: Monoclonal
Clone: 8G1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse