To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Synapse-Associated Protein 130
- Antigen Symbol:SAP130
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:23450
- Antigen Synonyms:SAP130SAP 130|splicing factor 3B subunit 3|SF3b130pre-mRNA splicing factor SF3b|130 kDa subunit|splicing factor 3b|Spliceosome-associated protein 130|subunit 3|KIAA0017splicing factor 3b|130kD|Pre-mRNA-splicing factor SF3b 130 kDa subunit|130kDa|STAF130|SF3B130|RSE1
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-VSKKLEDIRTRYAF corresponding to C-Terminus according to NP_036558.3.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102104-546
- Supplier no.:NB100-1077
Specifications
About this item
The SAP130 Antibody from Novus Biologicals is a goat polyclonal antibody to SAP130. This antibody reacts with human. The SAP130 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: SAP130
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human