To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Stromal Antigen 1
- Antigen Symbol:SA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Xenopus,Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10274
- Antigen Synonyms:cohesin subunit SA-1|SA1|nuclear protein stromal antigen 1|stromal antigen 1
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:A 36 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 5 and 6 (LocusLink ID 10274).
- Tested Applications:, Proximity Ligation Assay
- Purification:Immunogen affinity purified
- Cat. No.:102108-412
- Supplier no.:NB100-298
Specifications
About this item
The SA1 Antibody from Novus Biologicals is a goat polyclonal antibody to SA1. This antibody reacts with human, xenopus. The SA1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Proximity Ligation Assay.
Type: Primary
Antigen: SA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Xenopus