To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:F-Spondin
- Antigen Symbol:SPON1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:10418
- Antigen Synonyms:Vascular smooth muscle cell growth-promoting factor|f-spondin|spondin 1|(f-spondin) extracellular matrix protein|spondin-1|KIAA0762VSGP/F-spondin|extracellular matrix protein|VSGP|MGC10724
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.91 mg/ml
- Immunogen:Synthetic peptide, corresponding to amino acids 549-807 of SPON1.
- Purification:Immunogen affinity purified
- Cat. No.:102130-660
- Supplier no.:NB300-339
Specifications
About this item
The F-Spondin / SPON1 Antibody from Novus Biologicals is a chicken polyclonal antibody to F-Spondin / SPON1. This antibody reacts with human, mouse, rat. The F-Spondin / SPON1 Antibody has been validated for the following applications: Western Blot, ICC / IF (Negative).
Type: Primary
Antigen: SPON1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat