The disc filter holders are durable and of convenient size for filtration of virtually all laboratory samples.
- Durable
- Convenient size
- Corrosion-resistant
These disc filter holders are made of corrosion-resistant type 316 stainless steel that provide broad chemical resistance
They feature sanitary connections that are easy to use and suitable for pharmaceutical applications. Recommended uses include filtration of cell culture media, ophthalmics, pharmaceuticals, vitamins, process water, antibiotics, photoresists, etc. Materials of Construction- Body: Electropolished type 316 stainless steel; Rings: Viton; Legs: Type 316 stainless steel with copper threads; Knob Assembly: Aluminum with copper threads.
They can be used with a variety of aggressive chemicals, solvents, reagents, and fluids.
- Catalog No:
- 28145-269
- 28145-258
- Dimensions:
- Clearance between flange and benchtop: 29 cm (11.4")
- Inlet/Outlet Connections:
- Sanitary 3.8 cm (1.5") fittings
- Sanitary 3.8 cm (1.5") fittings
- Max. Operating Pressure:
- 6.9 (690 kPa, 100 psi)
- 6.9 (690 kPa, 100 psi)