White Fibrous Connective Tissue Slide
Catalog # 470176-958
Supplier: Avantor
Specifications
- Description:White fibrous connective tissue slide
- Environmentally Preferable:
- Cat. No.:470176-958
- Stain:Hematoxylin and Triosin
- Supplier no.:WARD470176-958
- Orientation:Longitudinal section
Specifications
About this item
Illustrates connective tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal tendon. Shows strands of cells in dense fiber bundles.