Skeletal Muscle Fibers Slide
470182-664
: Avantor
- Description:Skeletal Muscle Fibers
- Environmentally Preferable:
- Cat. No.:470182-664
- Stain:Periodic acid Schiff reaction
- Supplier no.:WARD470182-664
- Orientation:Section
Demonstrates Skeletal Muscle Fiber Characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. The tissue is stained to show “red” fibers and “white” fibers rich in glycogen. Brilliant magenta.