ContactUsLinkComponent
Lab-Aids® Osmosis And Diffusion Kit
Lab-Aids® Osmosis And Diffusion Kit
  470110-730
 :  Lab-Aids
undefined
Lab-Aids® Osmosis And Diffusion Kit
  470110-730
 :  Lab-Aids
 :  22
  • Description:
    Osmosis And Diffusion Kit
  • Environmentally Preferable:
  • No. of lab groups:
    30
  • Cat. No.:
    470110-730
  • Supplier no.:
    22
Observe the movement of water and solutes.
  • Observe diffusion through a membrane
  • Time required: one 50 minutes class period
  • Color changes help students visualize osmosis
Can studying the basic life process function of osmosis and diffusion stimulate your students' curiosity? It can with this hands-on lab experience! Students observe first hand the characteristics of a deferentially permeable membrane. Some substances will pass through the membrane and some won't. Some will actually pass through the membrane in both directions simultaneously. Simple color changes help students visualize this biological-physiological phenomenon.
 : See datasheet for contents information.