ContactUsLinkComponent
Elastic Cartilage, Mammal Slide
Elastic Cartilage, Mammal Slide
Catalog # 470177-580
Supplier:  Avantor
undefined
Elastic Cartilage, Mammal Slide
Catalog # 470177-580
Supplier:  Avantor
Specifications
  • Description:
    Elastic Cartilage, Mammal
  • Environmentally Preferable:
  • Cat. No.:
    470177-580
  • Stain:
    Verhoeff and Eosin
  • Supplier no.:
    WARD470177-580
  • Orientation:
    Section
Specifications
About this item
Examine an important structural tissue of the body
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal epiglottis. Stained to show elastic fibers.