Rotors for Allegra® X-30 and X-30R Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
Certificates
About this item
All rotors are for use with the Allegra® X-30 and X-30R Centrifuges.
- Swinging Bucket, Microplate, and Fixed-Angle Rotors Available
- Material: Aluminum
- Applications: Processing of Cell Cultures, Blood, Urine, and More
- Swinging Bucket and Fixed-Angle Rotors are BioCertified
- Microplate Rotor Maximum G-Force: 2721 g
Models SX4400, SX4250, and SX241.5 are swinging-bucket rotors that can centrifuge a wide range of tubes and bottle sizes.
SX4400 and SX4250 are supplied with four buckets, and SX241.5 is supplied with a lid. The rotor yokes, lids, and buckets are made with aluminum and black-anodized for corrosion protection.
Ordering information: Adapters are available to accommodate a variety of tubes and bottles. Adapters and BioSafe bucket covers are sold separately.