TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) for biotechnology
: VWR
: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride
:51805-45-9
:
:C₉H₁₆ClO₆P
:MFCD00145469
:286.65 g/mol
:Refrigerator
:3261
:8 II
:GHS05
- Environmentally Preferable:
- Purity (TLC):> 99%
- Solubility (50mg/ml, Water):pass