ContactUsLinkComponent
Filter Funnel, 47 mm, Stainless Steel, Cytiva (Formerly Pall Lab)
 :  Cytiva
undefined
Filter Funnel, 47 mm, Stainless Steel, Cytiva (Formerly Pall Lab)
 :  Cytiva
  • Catalog No:
  • 28144-958
  • 28144-903
  • Effective Filtration Area:
  • 9.6 cm²
  • 9.6 cm²
  • Sterilization:
  • Provided non-sterile. Autoclavable if desired at 121 - 123 °C (250 - 253 °F) at 1.0 bar (100 kPa, 15 psi) for 15 - 20 min.
  • Provided non-sterile. Autoclavable if desired at 121 - 123 °C (250 - 253 °F) at 1.0 bar (100 kPa, 15 psi) for 15 - 20 min.