Supplied at a 2X concentration and contains dATP, dTTP, dCTP, dGTP, and other buffer components and salts including Tris-HCl, MgCl₂, and KCl.
- Easy to use
- Versatile
- Provides excellent amplification under rapid conditions
- Storage temperature: −10 to −25 °C
- Shipping temperature: Dry ice
- Minimize freeze thaw to avoid loss of performance
SPRINTaq™ is a modified version of Empirical's standard Taq DNA polymerase, and the 2X MasterMix formula has been optimized for use in rapid PCR applications. Including endpoint PCR, and probe-based qPCR (e.g. TaqMan-like) applications. Just add template and primers with the MasterMix and the reaction is ready to go.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Environmentally Preferable: