Anti-CEP76 Rabbit Polyclonal Antibody
Catalog # 77690-490
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:CEP76
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q8TAP6
- Antigen Synonyms:HsT1705|CEP76_HUMAN|C18orf9|Centrosomal protein of 76 kDa|Cep76|Centrosomal protein 76kDa|CEP 76|FLJ12542|Chromosome 18 open reading frame 9
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Centrosomal protein of 76 kDa protein (241-550AA)
- Purification:>95%, Protein G purified
- Cat. No.:77690-490
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: CEP76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human