Anti-FKBP2 Rabbit Polyclonal Antibody
Catalog # 77687-665
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:FKBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P26885
- Antigen Synonyms:FK506 binding protein 2 (13kD)|Peptidyl-prolyl cis-trans isomerase FKBP2|Proline isomerase|T-cell|FKBP2|FK506-binding protein|FKBP13|FKBP2_HUMAN|13 kDa FKBP|Peptidyl prolyl cis trans isomerase|EC 5.2.1.8|FKBP-13|PPIase FKBP2|Rotamase|13-KD|FKBP-2|FK506 binding protein 2 13kDa|Immunophilin FKBP13|FKBP 2|Rapamycin binding protein|FKBP 13|13 kDa FK506-binding protein|PPIase|FK506-binding protein 2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP2 protein (22-142AA)
- Purification:Antigen affinity purified
- Cat. No.:77687-665
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200
Type:
Antigen: FKBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat