Arm-Evac® 50 Benchtop Fume Extraction System, PACE®
30138-404
: Avantor
- Description:Arm-Evac® 50 System, 115V
- Environmentally Preferable:
- Cat. No.:30138-404
- W×D×H:29×29×50 cm ( 13× 12½× 8½")
- Weight:6 kg (13 lbs.)
- Supplier no.:8889-0050
The portable Arm-Evac® 50 fume extractor provides wide-area fume extraction
Features include operator adjustable airflow to 90 cfm, quiet operation (54 dBA), one prefilter, and one general-purpose filter. Highly efficient source capture at two points is possible using the optional arm attachment (30138-392).