Barnstead/Thermolyne Explosion-Proof Hot Plate, Safe-T HP6, Thermo Scientific
33922-655
UL Listed
- Temperature Range:38 to 220°C
- Environmentally Preferable:
- Shipping Weight:4 kg (9.5 lbs.)
- Dimensions:15.6W x 15.6D cm (61/8 x 61/8") Top Plate, 17.8W x 8.3H x 19.4D cm
(7 x 31/4 x 75/8") Overall - Cat. No.:33922-655
- Electrical:120 V, 50/60 Hz, 600 W
- Supplier no.:HP11515B
Designed for Class I, Group D hazardous areas
Explosion-proof. “Limit” thermostat cuts off unit when temperature reaches 243°C (469.4°F). Sealed aluminum housing contains and protects controls. Cast aluminum top plate ensures maximum heat transfer. Corrosion-resistant steel case is easily cleaned and maintained. Integral thermostatic control maintains top plate temperature ±5.5°C (±9.9°F). Accommodates up to 11.3kg (25lb.) loads.
: Integral mounting lugs, connection for 1.27cm (1/2") conduit and 45.7cm (18") lead wire included for safe installation. Conduit installation required. For optional dial thermometer, see 33922-677.