About this item
Equally effective in both permanent and temporary applications, these modular enclosures are designed, engineered, and manufactured to offer great flexibility
Choose from three enclosure insert materials: STD-Clear, ESD-Clear, and LEXAN® plastic. The STD-Clear material is a fire-retardant standard grade vinyl with a thickness of 0.40mil. The ESD-Clear material is composed of a static dissipative vinyl with a thickness of 0.40mil. It has a surface resistivity of 1.8x1010ohm/sq. Clear LEXAN polycarbonate plastic is 3.2mm (1/8") thick.
The ISO Class 5 (FED-STD-209E Class 100/M3.5) rated fan-powered ceiling HEPA filter units are 99.99% efficient on particles of 0.3µm or larger.
This totally integrated wall panel and ceiling module system latches together for quick assembly with minimal use of tools. System provides strong, positive connection at walls, corners, and ceilings. Enclosures can be easily expanded or reconfigured to meet ever-changing demands. Wall panels, doors, and ceiling frames come preassembled. The anodized aluminum framing is available in powder-coat.