Order Entry
United States
ContactUsLinkComponent
Human IL10 ELISA Kit
Human IL10 ELISA Kit
Catalog # MSPP-SEA056HU
CAS Number:  
undefined
Human IL10 ELISA Kit
Catalog # MSPP-SEA056HU
Supplier Number:  SEA056HU
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL10
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 10 (IL10) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    7.8 - 500 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    2.8 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA056HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human IL10 (Interleukin 10). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 2.8 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Interleukin-10 is a small protein known as a cytokine that functions as an important regulator of the immune system.  Although IL-10 is known to have many different roles in the immune system, its two major activities include inhibition of cytokine production by macrophages and inhibition of the accessory functions of macrophages during T cell activation. The effects of these actions cause IL-10 to play mainly an anti-inflammatory role in the immune system. IL-10 was originally known as the cytokine synthesis inhibiting factor (CSIF), and the discovery of this protein was based on its biological activity. IL-10 is mainly produced by the Th2 subset of CD4+ helper cells.  However, it is also produced by some activated B cells, some Th1 cells, activated macrophages, and some nonhematopoietic sources.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.