To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:5-Hydroxytryptamine
- Antigen Symbol:5-HT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Size:200 µl
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:N-Methyl-Gamma|5HT|Serotonin
- Storage Buffer:0.01 M PBS, pH 7.4, containing 0.05% Proclin-300, 50% glycerol
- Molecular Weight:176.2 Da
- Storage Temperature:Store at 2...8 °C for frequent use,−20 °C for 24 months. Avoid repeated freeze-thaw cycles.
- Concentration:0.64 mg/ml (for reference only)
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:BSA conjugated 5-HT
- Purification:Antigen-specific affinity chromatography followed by Protein A affinity chromatography
- Cat. No.:MSPP-PAA808GE1
- Packaging:Vial
Specifications
About this item
Polyclonal Antibody to 5-Hydroxytryptamine (5-HT), derived from BSA conjugated 5-HT, is reactive with General species.
- High specificity, affinity and purity
- Extensive validation and batch-to-batch consistency
- Diverse range of conjugated and custom options
- Comprehensive validation data and documentation
- All in stock and fast global delivery
- 100% Quality and service satisfaction guarantee
Serotonin also called 5-hydroxytryptamine, is primarily found in the gastrointestinal (GI) tract and central nervous system (CNS) of animals. Approximately 80 percent of the human body's total serotonin is located in the enterochromaffin cells in the gut, where it is used to regulate intestinal movements. The remainder is synthesized in serotonergic neurons in the CNS where it has various functions, including the regulation of mood, appetite, sleep, muscle contraction, and some cognitive functions including memory and learning; and in blood platelets where it helps to regulate hemostasis and blood clotting. In addition to animals, serotonin is also found in fungi and plants. 5-hydroxytryptamine(5-HT) is an important neurotransmitter made by neurons in the central nervous system. These functions are mediated by a large number of receptors. After it is released by a neuron, 5-HT activates receptors located on nearby neurons.
Type: Primary
Antigen: 5-HT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: General species