Specifications
- Pack type:50 disks per cartridge
- Description:SPS Identification disks
- Environmentally Preferable:
- Cat. No.:76642-522
Specifications
About this item
HardyDisk™ SPS Identification disks evaluate the ability of an organism to grow in the presence of sodium polyanethol sulfonate.
- Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing
HardyDisk™ SPS Identification disks can be used to presumptively identify microorganisms including but not limited to Peptostreptococcus anaerobius, Gardnerella vaginalis, and Haemophilus ducreyi.