Fully automatic shoe cleaner features a built-in vacuum, disposable bags, and HEPA filter.
- Simple to operate
- Quiet operation
- Rugged design for long service life
- Large footprint for stability
- Speed control, brush motor
Five brushes for complete contamination removal.
Vacuum exhaust: 1.50×1.75" (ID×OD)
Main unit dimensions: 18×27×10.5" (W×L×H)
Overall height with handle: 40"
Weight: 50 lbs. (shipping weight: 58 lbs.)