To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:20 ml
- Environmentally Preferable:
- Storage Temperature:Room Temperature (Recommended in a cool and dark place, <15°C)
- Appearance:Purple to dark purple clear liquid to cloudy liquid.
- Cat. No.:TCB6104-20ML
Specifications
About this item
This product contains 8% SDS, 40% glycerol, 0.02% bromophenol blue and 0.25 M Tris HCl, pH approx. 6.8. Does not contain 2-Mercaptoethanol.
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation based on the Laemmli Method of SDS-PAGE system.
Preparation of non-reducing sample
A protein sample is mixed with the 4X SDS-PAGE sample buffer (3:1). If necessary, heating the mix solution and then cool down the tube at room temperature. The solution is ready for SDS-PAGE.
Preparation of reducing sample (reducing with 2-Mercaptoethanol)
Add 20 μl of 2-Mercaptoethanol per 80 μl of 4X SDS-PAGE sample buffer. Mix thoroughly. Then a protein sample is mixed with the sample buffer (3:1) and heating to 95 - 100 °C for 5 min. Cool down the tube at room temperature. The solution is ready for SDS-PAGE.
Reducing reagents: 2-Mercaptoethanol [for Electrophoresis], DL-Dithiothreitol [for Electrophoresis], etc.