Cut Resistant Gloves, A4 Cut Level
: Showa
New Product
ANSI cut level A4 the new XC810 combines an excellent grip, superb tactility, 18-gauge HPPE A4 cut-resistance and touchscreen compatibility for a multitude of industrial applications.
- HPPE reinforced
- Ergonomic
- Microporous nitrile coating
- Embossed grip
- Cut-resistant/lightweight
- Flexible/breathable
- Increased dexterity
- Seamless knit/form-fitting
- Abrasion-resistant
- Touch screen compatible
- Natural rubber latex free
Application in light assembly of oil-coated pieces, maintenance, mechanical and engineering, repair glass and windows, component assembly.
: ANSI/ISEA 105-2016 CUT.
: Pair per polybag: 12; Polybags per case: 6; Pair per case: 72.
- Catalog No:
- 77575-120
- 77575-122
- 77575-160
- 77575-138
- 77575-136
- Model:
- XC810
- XC810
- XC810
- XC810
- XC810
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE