Isolation Transformers, Tripp Lite
Supplier: Tripp Lite
Certificates
About this item
Provide line isolation, continuous noise filtering, and enhanced common mode surge suppression for any industrial, medical, office, or network environment
Units feature rugged all-metal housing in an upright tower format, two or more widely-spaced hospital-grade output receptacles, 1.8 m (6') power cord, hospital-grade input plug, circuit breaker overload protection, and lighted power switch.
Ideal for the protection of sensitive electronic equipment, these medical-grade isolation transformers provide an inexpensive alternative to dedicated circuits and site electrical upgrades.
Noise suppression rating: 35–65 dB, Total harmonic distortion: <1%, Efficiency: 96%
Certifications: UL (UL Series AGIS250YWN) and cUL-listed.
Ordering information: Two-year manufacturer's limited warranty.