- Description:ImmunoCult™ dendritic cell culture kit
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:MSPP-10985
For differentiation of human monocytes into dendritic cells.
Culture and differentiate human monocytes into dendritic cells (DCs) using the animal component-free (ACF) ImmunoCult™ dendritic cell culture kit.
For your convenience, the kit includes all the necessary components for you to generate active mature DCs from human monocytes in 7 days:
ImmunoCult™-ACF dendritic cell medium for the in vitro culture and differentiation of human monocytes into DCs.
ImmunoCult™-ACF dendritic cell differentiation Supplement to support the differentiation of immature DCs from human monocytes.
ImmunoCult™ dendritic cell maturation Supplement to support the maturation of immature human DCs.
For more information on protocols for culture and differentiation using ImmunoCult™ dendritic cell culture kit, please explore the Product Information Sheet (PIS).