ContactUsLinkComponent
Tissue-Tek® Slide Staining Set, Sakura
Tissue-Tek® Slide Staining Set, Sakura
Catalog # 25608-902
Supplier:  Sakura Finetek
undefined
Tissue-Tek® Slide Staining Set, Sakura
Catalog # 25608-902
Supplier:  Sakura Finetek
Supplier Number:  4451
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Slide Staining Set
  • Environmentally Preferable:
  • Cat. No.:
    25608-902
  • Supplier no.:
    4451
Specifications
About this item
Designed for histological or cytological staining
Provides simultaneous use of up to 12 solution wells, each deep enough to permit total immersion of 2.5x7.6cm (1x3") slides. Snap-locking cover on solvent-resistant metal frame prevents spills.
Ordering information: Complete set includes 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. Additional staining dishes and 24-slide holder with detachable handle are sold separately.