Specifications
- Depth:16"
- Width:4"
- Description:Wall corner filler
- Height:30"
- Material:Plastic laminate
- Environmentally Preferable:
- Cat. No.:33501-174
- Supplier no.:R1050301604
Specifications
About this item
Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
- Key component of case systems casework is the plastic laminate
- Case cabinetry gives adaptability necessary in today’s laboratories
A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware. The result — cabinets that are as attractive as they are durable.
The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves. Color: Beige.