ContactUsLinkComponent
Robotic Tips for Tecan Systems
  77507-646
New Product
Robotic Tips for Tecan Systems
  77507-646
 :  332007
New Product
  • Volume:
    50 µl
  • Environmentally Preferable:
  • Tip color:
    Clear
  • Packed:
    96 per pack, 4800 per case
  • Cat. No.:
    77507-646
  • Sterile:
    +
  • For use with:
    Tecan Systems
NEST robotic tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals.
  • Adopt medical grade polypropylene material, which is manufactured by precision mold, and the material batch is stable
  • DNase-and RNase-free, no heat source contamination
  • Electron beam sterilization
  • Manufacturing facility equipped with class 100000 clean-room, products packing under sterile condition
  • Unique process technology ensures that the inner surface of the tip is smooth and reduces liquid residue
  • Carry out structural drawing and measurement according to the original adapter to ensure that the product has good air tightness and adaptability
  • Online testing equipment is used to control the size and concentricity of products in the production line