Specifications
- Dimensions:14 cm x 3.7 m x 5.1 cm (5.5" x 12' x 2")
- Environmentally Preferable:
- Cat. No.:89065-410
- Supplier no.:PLR283
Specifications
About this item
Provides permanent or semi-permanent containment in high-traffic areas on concrete, asphalt or most sealed surfaces
Pre-drilled aluminum mounting strips with steel anchors secure berm to most surfaces; remove anchors to reposition, then seal with caulking (not included).
Create any configuration by combining with other Build-A-Berm components; ends overlap and can be sealed together for liquid-proof containment. When used with other 2"-high Build-A-Berm Barriers, orange color indicates area where drivers can enter and exit.
Constructed of 40-ounce, vinyl-coated fabric with an open-cell foam core; 2"-high berm compresses to approximately 1/4" when driven over; springs back to shape when vehicles pass. Durable enough to withstand indoor or outdoor traffic from trucks, loaded tow motors and other heavy vehicles with sufficient ground clearance.