(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Supplier: RPI
New Product
Certificates
About this item
Synonyms: (3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
CAS Number:82473-24-3
:
Formula:C₃₂H₅₈N₂O₈S
Molecular Weight:630.89 g/mol
Storage Temperature:Room Temperature
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Solubility:Clear and haze free
- Assay (HPLC):≥ 99.0 %
- Moisture (LOD):≤ 2.0 %
- Conductivity of solution:≤ 100 MICROMHOS/CM
- Identification:Pass Test
- Ignition Residue:≤ 0.1 %
- pH (1% Solution):5.0 - 8.0
- TLC N2 ASSAY:Conforms to reference
- Absorbance 260nm:≤ 0.05 nm
- Absorbance 350nm:≤ 0.01 nm
- BSE/TSE Free:BSE/TSE Free
- Country of Origin:Report