Order Entry
United States
ContactUsLinkComponent
Contamination Control Swabs, Foam Head, Rigid Tip, Foamtec International WCC
Contamination Control Swabs, Foam Head, Rigid Tip, Foamtec International WCC
  10810-752
undefined
Contamination Control Swabs, Foam Head, Rigid Tip, Foamtec International WCC
  10810-752
 :  HT1000

 

  • Environmentally Preferable:
  • Cat. No.:
    10810-752
  • Handle length:
    152.4 mm (6.000")
  • Head width:
    12.5 mm (0.500")
  • Head length:
    22.3 mm (0.880")
  • Handle width:
    6.4 mm (0.250")
  • Supplier no.:
    HT1000

 

 

Designed to meet demanding cleanroom applications

Can be used with alcohol, Freon* 113 fluorocarbon, acetone, and other solvents for assembly cleaning, and for epoxy application. Rigid-top swab features a 100ppi polyurethane head and a polypropylene handle.

 : Packaged in polybags.