ContactUsLinkComponent
Safepette® Dispenser
Catalog # 77497-230
Supplier:  Drummond
Safepette® Dispenser
Catalog # 77497-230
Supplier:  Drummond
Supplier Number:  SD2050
Specifications
  • Color:
    Blue
  • Description:
    Dispenser safepette manual
  • Environmentally Preferable:
  • Cat. No.:
    77497-230
Specifications
About this item
After centrifugation, a Safecap® tube is loaded into the Safepette® dispenser. The plunger is then depressed to move the self-sealing plug, which acts as a piston.
This eliminates the hazardous practice of scoring and snapping blood-filled tubes and the associated risk of exposure to potentially life-threatening blood-borne pathogens.