- Description:Dose Pouch
- Size:29.6 mL (1 oz.)
- Environmentally Preferable:
- Cat. No.:11212-752
- Supplier no.:684128
This one-step, acid phenolic cleaner and detergent is specifically formulated for use on hard, nonporous surfaces
The 29.6mL (1oz.) unit dose pouch (11212-752) is premeasured to ensure the correct dilution every time. Each 29.6mL (1oz.) pouch makes 7.6L (2gal.) of solution.
Gamma-irradiated to ensure sterility and double-bagged for ease of introduction into aseptic areas. Process LpH®st has a use dilution pH of 2.6–3.0 and is ideal for use in a disinfectant rotation program with Process Vesphene® IIst disinfectant cleaner (see 21899-876 series).