Support Rod Holder, Humboldt
Catalog # 60118-009
Supplier: Humboldt Manufacturing
Small Business Enterprise
Specifications
- Description:Support Rod Holder
- Environmentally Preferable:
- Cat. No.:60118-009
- Supplier no.:H-21160
Specifications
About this item
Consists of a heavy, cast-iron clamp protectively enameled, for fastening to edge of table, bench or shelf up to 38mm (11/2") thick
These support rod holder are made of steel.
Accommodates two 91/2mm (3/8") diameter support rods at right angles. With one 91/2mm (3/8") diameter by 508mm (20") plated steel rod.