ContactUsLinkComponent
Hemacytometer Cover Glass, Propper Manufacturing
Hemacytometer Cover Glass, Propper Manufacturing
Catalog # 48300-470
Supplier:  Propper
undefined
Hemacytometer Cover Glass, Propper Manufacturing
Catalog # 48300-470
Supplier:  Propper
Supplier Number:  08000100
Specifications
  • Description:
    Hemacytometer cover glasses, rectangular
  • L×W:
    26×20 mm
  • Environmentally Preferable:
  • Cat. No.:
    48300-470
  • Supplier no.:
    080001
Specifications
About this item
Propper hemacytometer cover glass is for your with red blood cell and platelet counting chambers.
  • Clear, non-fogging, non-corrosive glass
  • Chemical neutral glass will not disturb contents of slide
  • Highly resistant to breakage
Propper's Hemacytometer cover glass is designed to fit the exact distance between the cover glass and the Hemacytometer counting chamber. Precisely ground to ensure an exact distance between the cover glass and the surface of the ruled counting area. Cleaned and washed - ready to use.