Order Entry
United States
ContactUsLinkComponent
 
KingFisher™ Magnetic Particle Processor
  14230-360
undefined
KingFisher™ Magnetic Particle Processor
  14230-360
 :  5400000

 

  • Description:
    Particle processor
  • Environmentally Preferable:
  • Electrical:
    110 - 240 V
  • W×D×H:
    30×30×30 cm (11¹³/₁₆×11¹³/₁₆×11¹³/₁₆")
  • Weight:
    11.5 kg (25.3 lbs.)
  • Cat. No.:
    14230-360
  • Supplier no.:
    5400000

 

 

Purifies DNA, RNA, proteins, or cells.

Processor uses the principle of shifting magnetic particles through purification phases and produces no cross-contamination or reagent carryover. Purifies small-scale samples of 20 to 200 µl with up to 24 samples per run. Includes LCD display, fixed magnetic rods in 2×12 format, a magnetic particle processor with built-in software, and all essential plasticware.